Proteintech
Manufacturer Code:214001AP150UL
Catalog # 50-173-3407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPATA13 Fusion Protein Ag16101 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adenomatous polyposis coli stimulated exchange factor 2; APC-stimulated guanine nucleotide exchange factor 2; Asef2; ASEF2 SPATA13 spermatogenesis associated 13; Spermatogenesis-associated protein 13
Gene Aliases: ARHGEF29; ASEF2; C79050; ESTM11; mFLJ00298; SPATA13; X83327
UniProt ID: (Human) Q96N96, (Mouse) Q5DU57
Entrez Gene ID: (Human) 221178, (Rat) 305938, (Mouse) 219140
Molecular Function:
G-protein modulator
enzyme modulator
guanyl-nucleotide exchange factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.