Novus Biologicals
Manufacturer Code:NB10079793
Catalog # NB10079793
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human JNK/SAPK-Associated Protien using the numbering given in entry NP_003962.3 (GeneID 9043) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; Cancer/testis antigen 89; Cancer/testis antigen 89 c-Jun NH2-terminal kinase-associated leucine zipper protein C-Jun-amino-terminal kinase-interacting protein 4 CT89JIP4 FLJ14006 FLJ26141 FLJ34602 HLC4 HLC-6 HSS Human lung cancer oncogene 6 protein JLPPHETSYD1FLJ13450 JNK interacting protein JNK/SAPK-associated protein JNK-associated leucine-zipper protein JNK-interacting protein 4 KIAA0516JIP-4 lung cancer oncogene 4 MAPK8IP4 Max-binding protein MGC117291 MGC14967 MGC74461 Mitogen-activated protein kinase 8-interacting protein 4 PIG6 proliferation-inducing gene 6 Proliferation-inducing protein 6 Protein highly expressed in testis sperm associated antigen 9 Sperm surface protein Sperm-associated antigen 9 Sperm-specific protein Sunday driver 1; CT89; HLC-6; Human lung cancer oncogene 6 protein; JIP-4; JLP; JNK interacting protein; JNK-associated leucine-zipper protein; JNK/SAPK-associated protein; lung cancer oncogene 4; Max-binding protein; Mitogen-activated protein kinase 8-interacting protein 4; PHET; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; sperm associated antigen 9 variant 5; Sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1
Gene Aliases: CT89; HLC-6; HLC4; HLC6; HSS; JIP-4; JIP4; JLP; KIAA0516; MAPK8IP4; PHET; PIG6; SPAG9; SYD1
UniProt ID: (Human) O60271
Entrez Gene ID: (Human) 9043
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.