Abnova
Manufacturer Code:PAB2861
Catalog # 89-130-121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human SPAG9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SPAG9.
For Research Use Only
Protein Aliases: c-Jun NH2-terminal kinase-associated leucine zipper protein; C-Jun-amino-terminal kinase-interacting protein 4; Cancer/testis antigen 89; CT89; FLJ13450; FLJ14006; FLJ26141; FLJ34602; HLC-6; HLC4; Human lung cancer oncogene 6 protein; JIP-4; JLP; JNK interacting protein; JNK-associated leucine-zipper protein; JNK/SAPK-associated protein; KIAA0516; lung cancer oncogene 4; Max-binding protein; MGC117291; MGC14967; MGC74461; Mitogen-activated protein kinase 8-interacting protein 4; PHET; PIG6; proliferation-inducing gene 6; Proliferation-inducing protein 6; Protein highly expressed in testis; sperm associated antigen 9 variant 5; Sperm surface protein; Sperm-associated antigen 9; Sperm-specific protein; Sunday driver 1
Gene Aliases: CT89; HLC-6; HLC4; HLC6; HSS; JIP-4; JIP4; JLP; KIAA0516; MAPK8IP4; PHET; PIG6; SPAG9; SYD1
UniProt ID: (Human) O60271
Entrez Gene ID: (Human) 9043
Molecular Function:
enzyme modulator
kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.