Invitrogen
Manufacturer Code:PA597169
Catalog # PIPA597169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 220-420 of human SPAG8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SMP-1; sperm associated antigen 8-like protein; Sperm membrane protein 1; Sperm membrane protein BS-84; Sperm-associated antigen 8
Gene Aliases: MH-SPAG8; Spag8
UniProt ID: (Mouse) Q3V0Q6
Entrez Gene ID: (Rat) 362508, (Mouse) 433700
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.