Proteintech
Manufacturer Code:139151AP150UL
Catalog # 50-172-9087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPAG8 Fusion Protein Ag4910 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BS 84 HSD 1 hSMP 1 SMP 1 SMP1 SPAG3 SPAG8 sperm associated antigen 8 Sperm membrane protein 1 Sperm membrane protein BS 84; HSD-1; SMP-1; sperm associated antigen 8-like protein; Sperm membrane protein 1; Sperm membrane protein BS-84; Sperm-associated antigen 8; testicular tissue protein Li 177
Gene Aliases: BS-84; CILD28; CT142; HSD-1; hSMP-1; MH-SPAG8; SMP1; SPAG3; SPAG8
UniProt ID: (Human) Q99932, (Mouse) Q3V0Q6
Entrez Gene ID: (Human) 26206, (Mouse) 433700
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.