Novus Biologicals
Manufacturer Code:NBP15696020UL
Catalog # NBP15696020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SPAG4L(sperm associated antigen 4-like) The peptide sequence was selected from the N terminal of SPAG4L. Peptide sequence MPRSSRSPGDPGALLEDVAHNPRPRRIAQRGRNTSRMAEDTSPNMNDNIL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dJ726C3.1 MGC33594 Sad1 and UNC84 domain containing 5 SPAG4L Sperm-associated antigen 4-like protein SUN domain-containing protein 5 testis and spermatogenesis related gene 4 Testis and spermatogenesis-related gene 4 protein TSARG4sperm associated antigen 4-like; Sad1 and UNC84 domain-containing protein 5; Sperm-associated antigen 4-like protein; SUN domain-containing protein 5; testis and spermatogenesis related gene 4; Testis and spermatogenesis-related gene 4 protein
Gene Aliases: dJ726C3.1; SPAG4L; SUN5; TSARG4
UniProt ID: (Human) Q8TC36
Entrez Gene ID: (Human) 140732
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.