Proteintech
Manufacturer Code:174951AP150UL
Catalog # 50-173-1605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPAG4L Fusion Protein Ag11629 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sad1 and UNC84 domain-containing protein 5; SPAG4L SUN5 TSARG4; Sperm-associated antigen 4-like protein; SUN domain-containing protein 5; testis and spermatogenesis related gene 4; Testis and spermatogenesis-related gene 4 protein
Gene Aliases: 1700021O15Rik; dJ726C3.1; SPAG4L; SPAG4L-2; SRG4; SUN5; TSARG4
UniProt ID: (Human) Q8TC36, (Mouse) Q9DA32
Entrez Gene ID: (Human) 140732, (Mouse) 76407
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.