Proteintech
Manufacturer Code:197211AP150UL
Catalog # 50-173-2507
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acrosomal protein ACR55; cancer/testis antigen 127; Outer dense fiber-associated protein SPAG4; Sad1 and UNC84 domain containing 4; SPAG4 sperm associated antigen 4 SUN4; sperm antigen 4; sperm tail protein; Sperm-associated antigen 4 protein; SUN domain-containing protein 4
Gene Aliases: 1700041K21Rik; CT127; mKIAA4118; MNCb-0953; SPAG4; SUN4
UniProt ID: (Human) Q9NPE6, (Mouse) Q9JJF2
Entrez Gene ID: (Human) 6676, (Mouse) 245865
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
non-motor microtubule binding protein
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.