Invitrogen
Manufacturer Code:PA557995
Catalog # PIPA557995
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SPAG16 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Pf20 protein homolog; PF20 WDR29 WD repeat domain 29 pf20 protein homolog sperm-associated WD repeat protein sperm-associated antigen 16 protein; Sperm-associated antigen 16 protein; sperm-associated WD repeat protein; WD repeat domain 29
Gene Aliases: 4921511D23Rik; 4930524F24Rik; 4930585K05Rik; AV261009; PF20; RGD1565062; SPAG16; WDR29
UniProt ID: (Human) Q8N0X2, (Mouse) Q8K450
Entrez Gene ID: (Human) 79582, (Mouse) 66722, (Rat) 501158
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.