Novus Biologicals
Manufacturer Code:NBP297844AF647
Catalog # NB103712
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SPAG16 fragment. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp666P1710 FLJ22724 FLJ37717 Pf20 protein homolog PF20MGC87036 sperm associated antigen 16 sperm-associated antigen 16 protein sperm-associated WD repeat protein WD repeat domain 29 WDR29; Pf20 protein homolog; Sperm-associated antigen 16 protein; sperm-associated WD repeat protein; WD repeat domain 29
Gene Aliases: PF20; SPAG16; WDR29
UniProt ID: (Human) Q8N0X2
Entrez Gene ID: (Human) 79582
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.