Abnova
Manufacturer Code:PAB18625
Catalog # 89-120-182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at internal region of human SPAG16. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of SPAG16.
For Research Use Only
Protein Aliases: DKFZp666P1710; FLJ22724; FLJ37717; MGC87036; PF20; Pf20 protein homolog; Sperm-associated antigen 16 protein; sperm-associated WD repeat protein; WD repeat domain 29; WDR29
Gene Aliases: PF20; SPAG16; WDR29
UniProt ID: (Human) Q8N0X2
Entrez Gene ID: (Human) 79582
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.