Novus Biologicals
Manufacturer Code:NBP189136
Catalog # NBP189136
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TNNGIFQINSRRWCSNLTPNVPNVCRMYCSDLLNPNLKDTVICAMKITQEPQGLGYWEAWRHHCQGKDLTEWVDGCDF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 54; CT54; CT54ALLP17Cancer/testis antigen 54 LYC3Sperm protein reactive with ASA Lysozyme-like acrosomal sperm-specific secretory protein ALLP-17 Lysozyme-like protein 3 lysozyme-like sperm-specific secretory protein ALLP17 LYZL31700025M08Rik SLLP1MGC119058 sperm acrosome associated 3 sperm acrosome membrane-associated protein 3 sperm lysozyme like protein 1 Sperm lysozyme-like protein 1 Sperm protein reactive with antisperm antibodies SPRASA; lysozyme C; Lysozyme-like acrosomal sperm-specific secretory protein ALLP-17; Lysozyme-like protein 3; lysozyme-like sperm-specific secretory protein ALLP17; Sperm acrosome membrane-associated protein 3; Sperm acrosome membrane-associated protein 3, membrane form; Sperm acrosome membrane-associated protein 3, processed form; sperm lysozyme like protein 1; Sperm lysozyme-like protein 1; sperm lyzozyme-like acrosomal protein 1; Sperm protein reactive with antisperm antibodies; Sperm protein reactive with ASA
Gene Aliases: ALLP17; CT54; LYC3; LYZC; LYZL3; SLLP1; SPACA3; SPRASA; UNQ424/PRO862
UniProt ID: (Human) Q8IXA5
Entrez Gene ID: (Human) 124912
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.