Invitrogen
Manufacturer Code:PA524263
Catalog # PIPA524263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 46-74 amino acids from the N-terminal region of human SACA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SAMP32 sperm acrosomal membrane-associated protein 32 sperm acrosome membrane-associated protein 1; Sperm acrosomal membrane-associated protein 32; Sperm acrosome membrane-associated protein 1
Gene Aliases: SAMP32; SPACA1
UniProt ID: (Human) Q9HBV2
Entrez Gene ID: (Human) 81833
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.