Proteintech
Manufacturer Code:128291AP150UL
Catalog # 50-172-8282
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SPACA1 Fusion Protein Ag3549 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SAMP32 SPACA1 sperm acrosome associated 1; Sperm acrosomal membrane-associated protein 32; Sperm acrosome membrane-associated protein 1
Gene Aliases: 1700124L11Rik; 4930540L03Rik; RGD1560284; SAMP32; SPACA1
UniProt ID: (Human) Q9HBV2, (Mouse) Q9DA48
Entrez Gene ID: (Human) 81833, (Rat) 500432, (Mouse) 67652
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.