Invitrogen
Manufacturer Code:PA570643
Catalog # PIPA570643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human SP9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: buttonhead-like zinc finger transcription factor; buttonhead-like zinc finger transcription factor Sp9; Sp9 transcription factor homolog; Transcription factor Sp9; zinc finger protein 990; zinc finger transcription factor
Gene Aliases: RGD1559457; SP9; zgc:85876; ZNF990
UniProt ID: (Human) P0CG40, (Mouse) Q64HY3, (Zebrafish) Q6NW96
Entrez Gene ID: (Bovine) 517257, (Human) 100131390, (Mouse) 381373, (Rat) 366078, (Zebrafish) 405896
Molecular Function: DNA binding protein nucleic acid binding transcription cofactor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.