Invitrogen
Manufacturer Code:PA580057
Catalog # PIPA580057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human SP3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GC-binding transcription factor Sp3; specificity protein 3; SPR-2; trans-acting transcription factor 3; Transcription factor Sp3; Transcription factor Sp3 SPR-2 SP3
Gene Aliases: D130027J01Rik; SP3; SPR2
UniProt ID: (Human) Q02447, (Mouse) O70494
Entrez Gene ID: (Human) 6670, (Mouse) 20687, (Rat) 367846
Molecular Function:
DNA binding protein
nucleic acid binding
transcription cofactor
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.