Novus Biologicals
Manufacturer Code:NBP274306AF594
Catalog # NP274306A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C1 |
Immunogen | Recombinant protein fragment corresponding to amino acids 232-521 of human SP110 (NP_004501) produced in E.coli. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: FLJ22835 IFI41 IFI75 interferon-induced protein 41 30kD Interferon-induced protein 41/75 interferon-induced protein 75 52kD IPR1 phosphoprotein 41 phosphoprotein 75 SP110 nuclear body protein Speckled 110 kDa Transcriptional coactivator Sp110 VODI; interferon-induced protein 41, 30kD; Interferon-induced protein 41/75; interferon-induced protein 75, 52kD; phosphoprotein 41; phosphoprotein 75; Sp110 nuclear body protein; Speckled 110 kDa; Transcriptional coactivator Sp110
Gene Aliases: IFI41; IFI75; IPR1; SP110; VODI
UniProt ID: (Human) Q9HB58
Entrez Gene ID: (Human) 3431
Molecular Function:
DNA binding protein
HMG box transcription factor
chromatin/chromatin-binding protein
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.