Invitrogen
Manufacturer Code:PA576840
Catalog # PIPA576840
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SP100. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lysp100b Nuclear antigen Sp100 Nuclear autoantigen Sp 100 Nuclear autoantigen Sp-100 Nuclear autoantigen Sp100 Nuclear dot associated Sp100 protein Nuclear dot-associated Sp100 protein SP 100 SP100 SP100 HMG nuclear autoantigen SP100 nuclear antigen SP100&lowbarHUMAN Speckled 100 kDa; Nuclear autoantigen Sp-100; Nuclear dot-associated Sp100 protein; SP100-HMG nuclear autoantigen; Speckled 100 kDa
Gene Aliases: A430075G10Rik; lysp100b; SP100
UniProt ID: (Human) P23497, (Mouse) O35892
Entrez Gene ID: (Human) 6672, (Mouse) 20684
Molecular Function: DNA binding protein HMG box transcription factor chromatin/chromatin-binding protein nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.