Abnova
Manufacturer Code:H00006672B01
Catalog # 89-006-271
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SP100 (AAH11562.1 1 a.a. ~ 480 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human SP100 protein.
For Research Use Only
Protein Aliases: DKFZp686E07254; FLJ00340; FLJ34579; Nuclear autoantigen Sp-100; Nuclear dot-associated Sp100 protein; SP100-HMG nuclear autoantigen; Speckled 100 kDa
Gene Aliases: lysp100b; SP100
UniProt ID: (Human) P23497
Entrez Gene ID: (Human) 6672
Molecular Function:
DNA binding protein
HMG box transcription factor
chromatin/chromatin-binding protein
nucleic acid binding
signaling molecule
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.