Proteintech
Manufacturer Code:113771AP150UL
Catalog # 50-172-7107
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SP100 Fusion Protein Ag1939 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Lysp100b Nuclear autoantigen Sp 100 SP100 SP100 nuclear antigen Speckled 100 kDa; Nuclear autoantigen Sp-100; Nuclear dot-associated Sp100 protein; SP100-HMG nuclear autoantigen; Speckled 100 kDa
Gene Aliases: lysp100b; SP100
UniProt ID: (Human) P23497
Entrez Gene ID: (Human) 6672
Molecular Function: DNA binding protein HMG box transcription factor chromatin/chromatin-binding protein nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.