Novus Biologicals
Manufacturer Code:NB1001896
Catalog # NB1001896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence CSRIESPNENSNNSQ corresponding to internal region according to NP_612482.2 NP_003100.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Sp1 transcription factor specificity protein 1 transcription factor Sp1 TSFP1; specificity protein 1; Transcription factor Sp1
Gene Aliases: SP1; TSFP1
UniProt ID: (Human) P08047
Entrez Gene ID: (Human) 6667
Molecular Function: DNA binding protein nucleic acid binding transcription cofactor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.