R&D Systems
Manufacturer Code:AF3075SP
Catalog # AF3075SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human SOX9 Met1-Lys151 Accession # P48436 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: campomelic dysplasia autosomal sex-reversal CMD 1 CMD1 CMPD1 SRA1SRY (sex-determining region Y)-box 9 protein SRY (sex determining region Y)-box 9 SRY-related HMG-box gene 9 transcription factor SOX-9; SRY (sex determining region Y)-box9; SRY (sex-determining region Y)-box 9 protein; SRY box 9; SRY-related HMG-box, gene 9; Transcription factor SOX-9
Gene Aliases: CMD1; CMPD1; SOX9; SRA1; SRXX2; SRXY10
UniProt ID: (Human) P48436
Entrez Gene ID: (Human) 6662
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.