OriGene
Manufacturer Code:TA504949
Catalog # 50-167-5530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2A3 |
Immunogen | Full length human recombit protein of human SOX5 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: L-SOX5 L-SOX5B L-SOX5F LAMSHF; SRY (sex determining region Y)-box 5; SRY box 5; Transcription factor SOX-5
Gene Aliases: L-SOX5; L-SOX5B; L-SOX5F; LAMSHF; SOX5
UniProt ID: (Human) P35711
Entrez Gene ID: (Human) 6660
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.