Invitrogen
Manufacturer Code:PA540696
Catalog # PIPA540696
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SOX5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Sox5-BLM isoform; SRY (sex determining region Y)-box 5; SRY (sex determining region Y)-box 5 L-SOX5 MGC35153; SRY box 5; SRY-box 5; SRY-box containing gene 5; SRY-box containing gene 5-like 1; Transcription factor SOX-5
Gene Aliases: A730017D01Rik; AI528773; L-SOX5; L-SOX5B; L-SOX5F; LAMSHF; Sox-5; SOX5; Sox5l1
UniProt ID: (Human) P35711, (Rat) F1M8W4, (Mouse) P35710
Entrez Gene ID: (Rabbit) 100348988, (Human) 6660, (Horse) 100064442, (Bovine) 533829, (Dog) 486635, (Rat) 140587, (Mouse) 20678
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.