Novus Biologicals
Manufacturer Code:NBP279768100UG
Catalog # NB010553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SOX4/2540 |
Immunogen | Recombinant full-length human SOX4 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ecotropic viral integration site 16; ecotropic viral integration site 16 EVI16 SRY (sex determining region Y)-box 4 SRY-related HMG-box gene 4 transcription factor SOX-4; SRY (sex determining region Y)-box 4; SRY box 4; SRY-related HMG-box gene 4; Transcription factor SOX-4
Gene Aliases: EVI16; SOX4
UniProt ID: (Human) Q06945
Entrez Gene ID: (Human) 6659
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.