Invitrogen
Manufacturer Code:PA595290
Catalog # PIPA595290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human SOX4 identical to the related mouse sequence. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ecotropic viral integration site 16; SRY (sex determining region Y)-box 4; SRY box 4; SRY-box containing gene 4; SRY-related HMG-box gene 4; Transcription factor SOX-4; Transcription factor SOX-4 SOX4
Gene Aliases: AA682046; EVI16; Sox-4; SOX4
UniProt ID: (Human) Q06945, (Mouse) Q06831
Entrez Gene ID: (Human) 6659, (Mouse) 20677
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.