Invitrogen
Manufacturer Code:PA541442
Catalog # PIPA541442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SOX4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ecotropic viral integration site 16; SRY (sex determining region Y)-box 4; SRY (sex determining region Y)-box 4 EVI16; SRY box 4; SRY-box 4; SRY-box containing gene 4; SRY-related HMG-box gene 4; Transcription factor SOX-4
Gene Aliases: AA682046; EVI16; Sox-4; SOX4
UniProt ID: (Human) Q06945, (Mouse) Q06831
Entrez Gene ID: (Goat) 108633488, (Human) 6659, (Dog) 100687084, (Rabbit) 100353357, (Bovine) 768313, (Rat) 364712, (Guinea pig) 100733500, (Mouse) 20677
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.