Abnova
Manufacturer Code:H00006659A01
Catalog # 89-105-487
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 45 a.a. &bsim 136 a.a) partial recombinant protein with GST tag.,SOX4 (NP_003098 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SOX4.
For Research Use Only
Protein Aliases: ecotropic viral integration site 16; EVI16; SRY (sex determining region Y)-box 4; SRY box 4; SRY-related HMG-box gene 4; Transcription factor SOX-4
Gene Aliases: EVI16; SOX4
UniProt ID: (Human) Q06945
Entrez Gene ID: (Human) 6659
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.