Invitrogen
Manufacturer Code:MA531455
Catalog # PIMA531455
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT1876 |
Immunogen | Recombit protein encompassing a sequence within the center region of human SOX2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANOP3 antibody MCOPS3 antibody MGC2413 antibody SOX2 antibody transcription factor SOX2 antibody SRY-related HMG-box gene 2 antibody transcription factor SOX-2 antibody SRY (sex determining region Y)-box 2 antibody; sex determining region Y-box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY-box containing gene 2; SRY-related HMG-box gene 2; Transcription factor SOX-2; transcription factor SOX2
Gene Aliases: ANOP3; cb236; lcc; MCOPS3; RGD1565646; Sox-2; SOX2; soxp; wu:fb83g04; wu:fc14d07; ysb; zgc:65860; zgc:77389
UniProt ID: (Human) P48431, (Mouse) P48432, (Zebrafish) Q6P0E1
Entrez Gene ID: (Human) 6657, (Rat) 499593, (Mouse) 20674, (Zebrafish) 378723
Molecular Function: HMG box transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.