Novus Biologicals
Manufacturer Code:NBP229623SS
Catalog # NBP229623SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4G8 |
Immunogen | Full length recombinant human SOX2 (NCBI NP_003097). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ANOP3 MCOPS3 MGC2413 SRY (sex determining region Y)-box 2 SRY-related HMG-box gene 2 transcription factor SOX2 transcription factor SOX-2; sex determining region Y-box 2; SRY (sex determining region Y)-box 2; SRY box 2; SRY-related HMG-box gene 2; Transcription factor SOX-2; transcription factor SOX2
Gene Aliases: ANOP3; MCOPS3; SOX2
UniProt ID: (Human) P48431
Entrez Gene ID: (Human) 6657
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.