Abnova
Manufacturer Code:H00054345M01A
Catalog # 89-106-960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G12 |
Immunogen | SOX18 (NP_06088963 a.a. &bsim 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant SOX18.
For Research Use Only
Protein Aliases: HLTS; SRY (sex determining region Y)-box 18; SRY box 18; Transcription factor SOX-18
Gene Aliases: HLTRS; HLTS; SOX18
UniProt ID: (Human) P35713
Entrez Gene ID: (Human) 54345
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.