Invitrogen
Manufacturer Code:PA5115951
Catalog # PIPA5115951
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SOX18 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI385749 HLTRS HLTS Ra ragged Ragl SOX18 Sox-18 SRY (sex determining region Y)-box 18 SRY box 18 SRY-box 18 SRY-box containing gene 18 Sry-related HMG-box gene 18 transcription factor SOX-18; ragged; SRY (sex determining region Y)-box 18; SRY box 18; SRY-box containing gene 18; Sry-related HMG-box gene 18; Transcription factor SOX-18
Gene Aliases: AI385749; HLTRS; HLTS; Ra; Ragl; Sox-18; SOX18
UniProt ID: (Human) P35713, (Mouse) P43680
Entrez Gene ID: (Human) 54345, (Rat) 311723, (Mouse) 20672
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.