Novus Biologicals
Manufacturer Code:NBP241289
Catalog # NBP241289
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid peptide near the amino terminus of human SOX18. The immunogen is located within amino acids 40 - 90 of SOX18. Amino Acid Squence: ASPPSPQRSPPRSPEPG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HLTS SRY (sex determining region Y)-box 18 SRY-box 18 transcription factor SOX-18; SRY (sex determining region Y)-box 18; SRY box 18; Transcription factor SOX-18
Gene Aliases: HLTRS; HLTS; SOX18
UniProt ID: (Human) P35713
Entrez Gene ID: (Human) 54345
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.