OriGene
Manufacturer Code:TA600126
Catalog # 50-167-7211
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5G6 |
Immunogen | Recombit protein expressed in E.coli corresponding to amino acids 177-414 of human SOX17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: SRY (sex determining region Y)-box 17; SRY box 17; SRY-related HMG-box transcription factor SOX17; Transcription factor SOX-17; VUR3
Gene Aliases: SOX17; VUR3
UniProt ID: (Human) Q9H6I2
Entrez Gene ID: (Human) 64321
Molecular Function: HMG box transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.