Proteintech
Manufacturer Code:249031AP150UL
Catalog # 50-173-4608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SOX17 Fusion Protein Ag19157 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SOX17 Transcription factor SOX 17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box containing gene 17; SRY-related HMG-box transcription factor SOX17; Transcription factor SOX-17
Gene Aliases: Sox-17; SOX17; VUR3
UniProt ID: (Human) Q9H6I2, (Mouse) Q61473
Entrez Gene ID: (Human) 64321, (Mouse) 20671, (Rat) 312936
Molecular Function: HMG box transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.