Proteintech
Manufacturer Code:167251AP150UL
Catalog # 50-173-1141
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SOX15 Fusion Protein Ag10173 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein SOX 12 Protein SOX 15 Protein SOX 20 SOX12 SOX15 SOX20 SOX26 SOX27; Protein SOX-12; Protein SOX-15; Protein SOX-20; SRY (sex determining region Y)-box 15; SRY (sex determining region Y)-box 20; SRY box 15; SRY-box containing gene 15
Gene Aliases: Sox-15; SOX12; SOX15; SOX20; SOX26; SOX27
UniProt ID: (Human) O60248, (Mouse) P43267
Entrez Gene ID: (Human) 6665, (Mouse) 20670
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.