Proteintech
Manufacturer Code:189021AP150UL
Catalog # 50-173-2279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SOX13 Fusion Protein Ag13537 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ICA12 Islet cell antigen 12 Sox 13 SOX13 Transcription factor SOX 13; islet cell antibody 12; Islet cell antigen 12; mSox13; SRY (Sex determining region Y)-box 13; SRY box 13; SRY-box containing gene 13; SRY-related HMG-box gene 13; Transcription factor SOX-13; Type 1 diabetes autoantigen ICA12
Gene Aliases: AA407916; AW259412; AW540933; ICA12; Sox-13; SOX13
UniProt ID: (Human) Q9UN79, (Mouse) Q04891
Entrez Gene ID: (Human) 9580, (Mouse) 20668, (Rat) 289026
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.