Invitrogen
Manufacturer Code:PA537891
Catalog # PIPA537891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-SRLYYSFKNITKQHP from the internal region of SOX11 (aa309-323) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: SRY (sex-determining region Y)-box 11; SRY sex determining region Y-box 11 SRY sex-determining region Y-box 11 SRY-related HMG-box gene 11 transcription factor SOX-11; SRY-related HMG-box gene 11; Transcription factor SOX-11
Gene Aliases: MRD27; SOX11
UniProt ID: (Human) P35716
Entrez Gene ID: (Human) 6664
Molecular Function: HMG box transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.