Novus Biologicals
Manufacturer Code:NBP298872AF647
Catalog # NB106389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human SOX11. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SRY (sex determining region Y)-box 11 SRY (sex-determining region Y)-box 11 SRY-related HMG-box gene 11 transcription factor SOX-11; SRY (sex-determining region Y)-box 11; SRY-related HMG-box gene 11; Transcription factor SOX-11
Gene Aliases: MRD27; SOX11
UniProt ID: (Human) P35716
Entrez Gene ID: (Human) 6664
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.