Proteintech
Manufacturer Code:667861IG150UL
Catalog # 50-173-7187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D2C8 |
Immunogen | SOX10 Fusion Protein Ag23191 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DOM SOX10 Transcription factor SOX 10 WS2E WS4; dominant megacolon, mouse, human homolog of; SRY (sex determining region Y)-box 10; SRY box 10; SRY-related HMG-box gene 10; Transcription factor SOX-10
Gene Aliases: DOM; PCWH; SOX10; WS2E; WS4; WS4C
UniProt ID: (Human) P56693
Entrez Gene ID: (Human) 6663
Molecular Function:
HMG box transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.