Novus Biologicals
Manufacturer Code:NBP211912AF750
Catalog # NP211912A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0556-9N13 |
Immunogen | Human Sclerostin |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Sclerostin; sclerostin VBCHsclerosteosis
Gene Aliases: CDD; DAND6; SOST; SOST1; UNQ2976/PRO7455/PRO7476; VBCH
UniProt ID: (Human) Q9BQB4
Entrez Gene ID: (Human) 50964
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.