Novus Biologicals
Manufacturer Code:NB11068800AF532
Catalog # N1106880A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | SOS-01 |
Immunogen | Peptide corresponding to amino acids THPSMHRDGPPLLENAHSS of human Sos protein (NM_005633.3) |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GF1GGF1 GINGF guanine nucleotide exchange factor HGFhereditary 1 NS4 son of sevenless homolog 1 son of sevenless homolog 1 (Drosophila) SOS-1; gingival fibromatosis, hereditary, 1; guanine nucleotide exchange factor; Son of sevenless homolog 1; SOS-1
Gene Aliases: GF1; GGF1; GINGF; HGF; NS4; SOS1
UniProt ID: (Human) Q07889
Entrez Gene ID: (Human) 6654
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.