Abnova
Manufacturer Code:MAB5560
Catalog # 89-112-299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AF3F11 |
Immunogen | Recombinant His/ABD fusion protein corresponding to human SOS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant SOS1.
For Research Use Only
Protein Aliases: GF1; GGF1; GINGF; gingival fibromatosis, hereditary, 1; guanine nucleotide exchange factor; HGF; NS4; Son of sevenless homolog 1; SOS-1
Gene Aliases: GF1; GGF1; GINGF; HGF; NS4; SOS1
UniProt ID: (Human) Q07889
Entrez Gene ID: (Human) 6654
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.