Invitrogen
Manufacturer Code:PA5105249
Catalog # PIPA5105249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SOS1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4430401P03Rik 9630010N06 AI449023 G GINGF GF1 GGF1 GINGF gingival fibromatosis gingival fibromatosis hereditary 1 guanine nucleotide exchange factor guanine nucleotide releasing factor HGF NS4 ras guanine exchange factor son of sevenless homolog 1 son of sevenless homolog 1 (Drosophila) son-of-sevenless 1 SOS Ras/Rac guanine nucleotide exchange factor 1 Sos1 SOS-1; gingival fibromatosis, hereditary, 1; guanine nucleotide exchange factor; guanine nucleotide releasing factor; ras guanine exchange factor; Son of sevenless homolog 1; son-of-sevenless 1; SOS-1
Gene Aliases: 4430401P03Rik; 9630010N06; AI449023; GF1; GGF1; GINGF; HGF; NS4; SOS1
UniProt ID: (Human) Q07889, (Mouse) Q62245
Entrez Gene ID: (Human) 6654, (Rat) 313845, (Mouse) 20662
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.