Abnova
Manufacturer Code:PAB0734
Catalog # 89-114-381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to extracellular region of human SORT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with deionized water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant SORT1.
For Research Use Only
Protein Aliases: 100 kDa NT receptor; Glycoprotein 95; Gp95; Neurotensin receptor 3; NT3; Sortilin
Gene Aliases: Gp95; LDLCQ6; NT3; NTR3; SORT1
UniProt ID: (Human) Q99523
Entrez Gene ID: (Human) 6272
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.