Invitrogen
Manufacturer Code:PA517941
Catalog # PIPA517941
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (TGFSDDVPMVIA) corresponding to the C-terminus amino acids of SORL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Gp250; LDLR relative with 11 ligand-binding repeats; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; mSorLA; SORL1 LR11 sortilin-related receptor L(DLR class) A repeats-containing LRP9 SORLA gp250 SorLA-1 mosaic protein LR11 C11orf32 FLJ21930 FLJ39258 sortilin-related receptor containing LDLR class A repeats; SorLA-1; Sortilin-related receptor; Sorting protein-related receptor containing LDLR class A repeats
Gene Aliases: 2900010L19Rik; AI596264; AW261561; gp250; LR11; mSorLA; Sorl1; SorLA
UniProt ID: (Mouse) O88307
Entrez Gene ID: (Mouse) 20660
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.