Invitrogen
Manufacturer Code:PA5115949
Catalog # PIPA5115949
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SorLA corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2900010L19Rik AI596264 AW261561 C11orf32 FLJ21930 FLJ39258 Gp250 LDLR relative with 11 ligand-binding repeats Low-density lipoprotein receptor relative with 11 ligand-binding repeats Lr11 LRP9 mosaic protein LR11 mSorLA RGD619914 SORL1 SorLA SorLA-1 sortilin related receptor 1 sortilin-related receptor sortilin-related receptor L(DLR class) A repeats containing sortilin-related receptor LDLR class A repeats-containing sorting protein-related receptor containing LDLR class A repeats; Gp250; LDLR relative with 11 ligand-binding repeats; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; mosaic protein LR11; mSorLA; SorLA; SorLA-1; Sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; Sorting protein-related receptor containing LDLR class A repeats
Gene Aliases: 2900010L19Rik; AI596264; AW261561; C11orf32; gp250; LR11; LRP9; mSorLA; RGD619914; SORL1; SORLA; SorLA-1
UniProt ID: (Human) Q92673, (Rat) P0DSP1, (Mouse) O88307
Entrez Gene ID: (Human) 6653, (Rat) 300652, (Mouse) 20660
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.