Abnova
Manufacturer Code:H00006653A01
Catalog # 89-003-086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | SORL1 (NP_003096 82 a.a. ~ 181 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SORL1.
For Research Use Only
Protein Aliases: C11orf32; FLJ21930; FLJ39258; gp250; LDLR relative with 11 ligand-binding repeats; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; LR11; LRP9; mosaic protein LR11; SORLA; SorLA-1; Sortilin-related receptor; sortilin-related receptor, L(DLR class) A repeats containing; Sorting protein-related receptor containing LDLR class A repeats
Gene Aliases: C11orf32; gp250; LR11; LRP9; SORL1; SORLA; SorLA-1
UniProt ID: (Human) Q92673
Entrez Gene ID: (Human) 6653
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.