Novus Biologicals
Manufacturer Code:NB10056696
Catalog # NB10056696
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This polyclonal antibody was raised against a peptide corresponding to amino acids 443-457 of human SODD protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 4; BAG-4; BAG-4 BAG-family molecular chaperone regulator-4 Bcl-2-associated athanogene 4 BCL2-associated athanogene 4 Silencer of death domains SODDBAG family molecular chaperone regulator 4; BAG-family molecular chaperone regulator-4; Bcl-2-associated athanogene 4; BCL2-associated athanogene 4; Silencer of death domains
Gene Aliases: BAG-4; BAG4; SODD
UniProt ID: (Human) O95429
Entrez Gene ID: (Human) 9530
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.