R&D Systems
Manufacturer Code:AF4349
Catalog # AF4349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse SODD/BAG4 Met1-Leu457 Accession # Q8CI61 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BAG family molecular chaperone regulator 4; BAG-4; BAG-family molecular chaperone regulator-4; BAG4 BAG-4 BAG-family molecular chaperone regulator-4 Bcl-2-associated athanogene 4 BCL2-associated athanogene 4 Silencer of death domains SODDBAG family molecular chaperone regulator 4; Bcl-2-associated athanogene 4; BCL2-associated athanogene 4; Silencer of death domains
Gene Aliases: BAG-4; BAG4; SODD
UniProt ID: (Human) O95429
Entrez Gene ID: (Human) 9530
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.